Rotavapor R-300 System B-305, SJ24/40, C, P+G, I-300P, V-300, 230V. Inc. V-300 pump, I-300 int. w/o chiller. Lift System: Manual Hea ting Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Cold trap, P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11SR300152CP01
Supplier: BUCHI
Frequently Bought Together