Rotavapor R-300 B-305, SJ29/32, C, P+G, I-300, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Cold trap, P+G coating, int.: I-300, Woulff bottle 1 * 1 items
Codice catalogo BUCH11R300151C121
Supplier: BUCHI
Frequently Bought Together