Order Entry
Italy
ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, C, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Cold trap, P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300101C151
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, C, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Cold trap, P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300101C151
Supplier:  BUCHI
Frequently Bought Together