Order Entry
Italy
ContactUsLinkComponent
Rotavapor R-300 no bath, SJ24/40, C, P+G, I-300, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Cold trap, P+G c oating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300102C121
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ24/40, C, P+G, I-300, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Cold trap, P+G c oating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300102C121
Supplier:  BUCHI
Frequently Bought Together