Rotavapor R-300 no bath, SJ29/32, S, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical re flux (S) P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
Codice catalogo BUCH11R300101S151
Supplier: BUCHI
Frequently Bought Together