Order Entry
Italy
ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, C, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: C old trap, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300112C051
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, C, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: C old trap, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300112C051
Supplier:  BUCHI
Frequently Bought Together