Order Entry
Italy
ContactUsLinkComponent
testo 526-2 - High precision differential pressure measuring instrument. Measuring range 0 to 2000 hPa (accuracy 0.05 % of f.v.). Delivery incl. fast coupling connections, battery and calibration protocol 1 * 1 items
Catalog # TEST0560.5281
Nome del fornitore:  TESTO
testo 526-2 - High precision differential pressure measuring instrument. Measuring range 0 to 2000 hPa (accuracy 0.05 % of f.v.). Delivery incl. fast coupling connections, battery and calibration protocol 1 * 1 items
Catalog # TEST0560.5281
Nome del fornitore:  TESTO
Informazioni correlate
Frequently Bought Together