VWR® Rota® Duo, pH indicator papers
7,0 - 10,0, 7,0 · 7,3 · 7,6 · 7,9 · 8,2 · 8,5 · 8,8 · 9,1 · 9,4 · 9,7 · 10,0
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Duo indicator papers show two different colours for each pH value.
- Easy to use for qualitatively accurate readings
- Produced from high quality paper to give good colour matching of paper to printed calibration scale
- Narrow range papers with graduation intervals of 0.3 pH units on the colour scale
- Wide range papers with graduation intervals up to 1 pH unit on the colour scale
Delivery information: Supplied in plactic reels that ensure protection and long-term stability. Reel of 5 m length, width: 10 mm
Documents
Certificates
Search by Lot Number85406_600
Images 360
VWR Indicator Papers and Strips.pdf
Product Literature
Specifications
- pH gradation:7,0 · 7,3 · 7,6 · 7,9 · 8,2 · 8,5 · 8,8 · 9,1 · 9,4 · 9,7 · 10,0
- pH range:7,0 - 10,0
- Seq:14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



