Order Entry
Italy
ContactUsLinkComponent
Temperature restriction (for UN/UF/UNplus/UFplus and models UF TS) Temperatures: +60, +70, +80, +95, +100, +120, +160, +180, +200, + 220 or +250 °C (Please, indicate upon ordering) 1 * 1 items
Catalog # MEMMUF160TSA8
Supplier:  MEMMERT
Temperature restriction (for UN/UF/UNplus/UFplus and models UF TS) Temperatures: +60, +70, +80, +95, +100, +120, +160, +180, +200, + 220 or +250 °C (Please, indicate upon ordering) 1 * 1 items
Catalog # MEMMUF160TSA8
Supplier:  MEMMERT
Frequently Bought Together