Order Entry
Italy
ContactUsLinkComponent
Rotator RT10, mixing angle 0-90, speed 5-60 rpm, digital controller, LCD display, timer 99 hr., 59 min., 40 W, 200 x 250 x 260 mm, Weight: 8,5 kg, 230 V, 50/60 Hz, * Unit only, without accessories * 1 * 1 items
Catalog # WITGDH.WRT00010
Supplier:  witeg Labortechnik
Rotator RT10, mixing angle 0-90, speed 5-60 rpm, digital controller, LCD display, timer 99 hr., 59 min., 40 W, 200 x 250 x 260 mm, Weight: 8,5 kg, 230 V, 50/60 Hz, * Unit only, without accessories * 1 * 1 items
Catalog # WITGDH.WRT00010
Supplier:  witeg Labortechnik
Frequently Bought Together