Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Ghost Dye™ Red 710

Red 710, 100 Tests
Catalog # 13-0871-T100
Supplier:   Tonbo Biosciences
Ghost Dye™ Red 710
Catalog # 13-0871-T100
Supplier:  Tonbo Biosciences
CAS Number:  
Ghost Dye™ Red 710
Ghost Dye™ Red 710

Red 710, 100 Tests

CAS Number:  
Price
49,90 €
Log in for availability
1 * 100 Tests
Product Details & Documents

Ghost Dye Red 710 is an amine reactive viability dye, that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications.

This dye irreversibly binds free amines available on the cell surface, as well as intracellular free amines exposed in cells with compromised cell membranes.

Necrotic cells with compromised membranes will react with significantly more Ghost Dye Red 710 dye than viable cells in the same sample and therefore exhibit much greater fluorescence intensity, allowing exclusion of these cells from analysis.

Specifications

Specifications

  • Pk:
    100 Tests
  • Description:
    Red 710
  • Cat. No.:
    13-0871-T100
  • Storage temperature:
    At ‒20 °C protected from light and moisture
  • Application:
    Flow cytometry

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.