Methyl 4-(1-hydroxyethyl)benzoate 90% tech.
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Methyl 4-(1-hydroxyethyl)benzoate 90% tech.
Catalog # ACRO424750050
Supplier: Thermo Fisher Scientific
CAS Number: 79322-76-2
Jump to:
Product Details & Documents
CAS No.: 79322-76-2
Synonyms: 1-[4-(Methoxycarbonyl)phenyl]ethan-1-ol, 4-(Methoxycarbonyl)-α-methylbenzyl alcohol
- CAS Number:79322-76-2
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD00216476
- Molecular Weight:180.203 g/mol
- Boiling Pt:148…152 °C (1 mmHg)
- Flash Pt:>93 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5300 to 1.5360 (20°C, 589 nm)
- GC:≥88 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



