Order Entry
Italy
Orders LinkContactUsLinkComponent
Ninhydrin ACS

Ninhydrin ACS

Glass bottle, 1 kg
Catalog # ACRO415720010
Irritant
Ninhydrin ACS
Catalog # ACRO415720010
CAS Number:  485-47-2
Ninhydrin ACS
Ninhydrin ACS

Glass bottle, 1 kg

CAS Number:  485-47-2
Price
1 450,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250 °C (dec.)
  • Density:0,862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light brown Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    Passes test
  • Solubility:
    Passes test
  • Identification:
    Passes test
  • Sensitivity:
    (to amino acids) Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

Specifications