Soda lime, granular, pure indicator, with indicator
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
Soda lime, granular, pure indicator, with indicator
Catalog # ACRO388352500
Supplier: Thermo Fisher Scientific
CAS Number: 8006-28-8
Jump to:
Product Details & Documents
With indicator white to violet
- CAS Number:8006-28-8
- MDL Number:MFCD00134094
- Storage Temperature:Ambient
- ADR:8,III
Documents
ACRO38835
SDS
Specifications
Specification Test Results
- Appearance:White to beige to light grey Granules
- Loss on drying:≤20 % (10 g, 200°C, 2 h)
- CO2 absorption capacity:≥19.0 %
Specifications
- Pack type:Glass bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



