Soda lime, granular, pure indicator, with indicator
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
Soda lime, granular, pure indicator, with indicator
Catalog # ACRO388350500
Supplier: Thermo Fisher Scientific
CAS Number: 8006-28-8
Jump to:
Product Details & Documents
With indicator white to violet
- CAS Number:8006-28-8
- MDL Number:MFCD00134094
- Storage Temperature:Ambient
- ADR:8,III
Documents
ACRO38835
SDS
Specifications
Specification Test Results
- Appearance:White to beige to light grey Granules
- Loss on drying:≤20 % (10 g, 200°C, 2 h)
- CO2 absorption capacity:≥19.0 %
Specifications
- Pack type:Glass bottle
- Pk:50 g



