Order Entry
Italy
Orders LinkContactUsLinkComponent
Polysorbate 40

Polysorbate 40

Glass bottle, 2,5 L
Catalog # ACRO334140025
Polysorbate 40
Catalog # ACRO334140025
CAS Number:  9005-66-7
Polysorbate 40
Polysorbate 40

Glass bottle, 2,5 L

CAS Number:  9005-66-7
Price
147,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:~ 0,1 °C
  • Density:1.05 g/cm³
  • Flash Pt:>200 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear to hazy light yellow to dark yellow Viscous liquid or paste
  • Infrared spectrum:
    Conforms
  • Acid value:
    ≤3 mg KOH/g
  • Hydroxyl value:
    89 to 105 mg KOH/g
  • Saponification value:
    41 to 55 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.