Kojic acid 99%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
- CAS Number:501-30-4
- Formula:C₆H₆O₄
- MDL Number:MFCD00006580
- Molecular Weight:142.11 g/mol
- Merck Index:13,05334
- Boiling Pt:323 °C (1013 hPa)
- Melting Pt:151…156 °C
- Density:1,58 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-922-4
Specifications
Specification Test Results
- Appearance:White to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:151 to 156 °C
- Titration with NaOH:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:500 g



