β-Alanine 99%
Plastic drum, 10 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
- CAS Number:107-95-9
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008200
- Molecular Weight:89.09 g/mol
- Merck Index:13,00204
- Boiling Pt:201 °C (1013 hPa)
- Melting Pt:197…202 °C (dec.)
- Density:1,44 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-536-5
Documents
ACRO16379
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 3 h)
- Sulfated ash:≤0.2 %
- pH:6.5 to 7.5 (10 % w/v in water)
- Titration with HClO4:≥98.5 % (on dry substance)
- Chloride (Cl):≤400 ppm
- Sulfate (SO4):≤480 ppm
- Ammonium (NH4):≤1000 ppm
- Iron (Fe):≤30 ppm
- Arsenic oxide (As2O3):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:(TLC) Passes test (not detectable on 10 µg spotting)
Specifications
- Pack type:Plastic drum
- Pk:10 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



