Selenium dioxide 99.8% (trace metals basis)
Glass bottle, 2,5 kg
Nome del fornitore: Thermo Fisher Scientific
Selenium dioxide 99.8% (trace metals basis)
Codice catalogo ACRO200000025
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 7446-08-4
Jump to:
Product Details & Documents
- CAS Number:7446-08-4
- Formula:SeO₂
- MDL Number:MFCD00003562
- Molecular Weight:110.96 g/mol
- Merck Index:13,08508
- Melting Pt:315 °C (subl.)
- Density:3,95 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-194-7
- UN:3283
- ADR:6.1,II
Documents
ACRO20000
SDS
Specifications
Specification Test Results
- Appearance:White to light beige Powder
- Assay:70 to 71.5 % Se
- Total trace metal impurities:≤0.2 %
- Arsenic (As):≤20 ppm
- Cadmium (Cd):≤5 ppm
- Copper (Cu):≤10 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤50 ppm
- Mercury (Hg):≤5 ppm
- Nickel (Ni):≤10 ppm
- Tellurium (Te):≤500 ppm
- Insoluble matter:≤0.2 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



