1-Hexene 97%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:592-41-6
- Formula:C₆H₁₂
- MDL Number:MFCD00009505
- Molecular Weight:84.16 g/mol
- Boiling Pt:62…65 °C (760 mmHg)
- Melting Pt:–140 °C
- Density:0.678 g/cm³
- Flash Pt:–26 °C
- Storage Temperature:Ambient
- EINECS:209-753-1
- UN:2370
- ADR:3,II
Documents
Certificates
Search by Lot NumberACRO21321
SDS
Thermo Scientific Stockroom Essentials
Product Literature
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3867 to 1.3887 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



