1-Octadecanol 95%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
- CAS Number:112-92-5
- Formula:H₃C(CH₂)₁₆CH₂OH
- MDL Number:MFCD00002823
- Molecular Weight:270.5 g/mol
- Merck Index:12,08960
- Boiling Pt:210 °C (15 mmHg)
- Melting Pt:55…58 °C
- Density:0,805 to 0.815 g/cm³ (60 °C)
- Flash Pt:185 °C
- Storage Temperature:Ambient
- EINECS:204-017-6
Documents
Specifications
Specification Test Results
- Appearance:White Powder or flakes or pellets
- Infrared spectrum:Conforms
- Melting point:55 to 60 °C
- GC:≥94 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



