2-Naphthaldehyde 98%
Glass bottle, 100 g
: β-Naphthaldehyde
- :66-99-9
- :C₁₁H₈O
- :MFCD00004094
- :156.18 g/mol
- :160 °C (19 mmHg)
- :58…62 °C
- :1,0775 g/cm³ (99 °C)
- :>100 °C
- :Ambient
- :200-640-2
- :3335
- :9,III
- Appearance:Off-white to yellow to light beige Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:58 to 62 °C
- GC:≥97.5 %
- Pack type:Glass bottle
- Pk:100 g



