Order Entry
Italy
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate 99%

Methyl-4-hydroxybenzoate 99%

Plastic drum, 10 kg
Catalog # ACRO126960100
Irritant
Methyl-4-hydroxybenzoate 99%
Catalog # ACRO126960100
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate 99%
Methyl-4-hydroxybenzoate 99%

Plastic drum, 10 kg

CAS Number:  99-76-3
Price
1 030,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,06129
  • Boiling Pt:298.6 °C (760 mmHg)
  • Melting Pt:125…128 °C
  • Density:1,1208 g/cm³ (137 °C)
  • Storage Temperature:Ambient
  • EINECS:202-785-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    125 to 128 °C
  • HPLC:
    ≥98.5 %
  • Residue after ignition:
    ≤0.1 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications