Malonic acid 99%
Plastic bottle, 5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Merck Index:13,05732
- Boiling Pt:387 °C (1013 hPa)
- Melting Pt:130…135 °C (dec.)
- Density:1,669 g/cm³ (20 °C)
- Flash Pt:172 °C
- Storage Temperature:Ambient
- EINECS:205-503-0
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:133 to 136 °C
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤500 ppm
Specifications
- Pack type:Plastic bottle
- Pk:5 kg



