Sanger's reagent 98%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
- CAS Number:70-34-8
- Formula:C₆H₃FN₂O₄
- MDL Number:MFCD00007056
- Molecular Weight:186.1 g/mol
- Merck Index:13,04199
- Boiling Pt:296 °C
- Melting Pt:23…26 °C
- Density:1.48 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:200-734-3
- UN:2811
- ADR:6.1,II
Documents
ACRO11703
SDS
Specifications
Specification Test Results
- Appearance:Yellow to brown Liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.5675 to 1.5705 (20°C, 589 nm)
- GC:≥97.5 %
- Residue after ignition:≤0.05 %
- Suitability:(for precip. of Fe, etc.) Passes test
Specifications
- Pack type:Glass bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



