Dichloroacetic acid (DCA) 99+%
Glass bottle, 2,5 L
: DCA Deblock, DCA
- :79-43-6
- :C₂H₂Cl₂O₂
- :MFCD00004223
- :128.94 g/mol
- :13,03075
- :194 °C (760 mmHg)
- :9…11 °C
- :1.56 g/cm³
- :>112 °C
- :Ambient
- :201-207-0
- :1764
- :8,II
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- Color scale:≤100 APHA
- Titration with NaOH:99.0 to 101.0 %
- Pack type:Glass bottle
- Pk:2,5 L



