Order Entry
Italy
Orders LinkContactUsLinkComponent
1,6-Hexanediol 97%

1,6-Hexanediol 97%

Plastic drum, 10 kg
Catalog # ACRO120650100
1,6-Hexanediol 97%
Catalog # ACRO120650100
CAS Number:  629-11-8
1,6-Hexanediol 97%
1,6-Hexanediol 97%

Plastic drum, 10 kg

CAS Number:  629-11-8
Price
420,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Hexamethylene glycol, 1,6-Dihydroxy hexane
  • CAS Number:629-11-8
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00002985
  • Molecular Weight:118.18 g/mol
  • Merck Index:13,04709
  • Boiling Pt:253…260 °C (760 mmHg)
  • Melting Pt:40…43 °C
  • Density:0.96 g/cm³
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:211-074-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless to white to light yellow Solidified melt or liquid
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 44 °C
  • Water:
    ≤0.3 % (K.F.)
  • GC:
    ≥96.0 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.