1,7-Heptanediol 95%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,7-Dihydroxy heptane
- CAS Number:629-30-1
- Formula:C₇H₁₆O₂
- MDL Number:MFCD00002987
- Molecular Weight:132.2 g/mol
- Boiling Pt:259 °C (760 mmHg)
- Melting Pt:17…19 °C
- Density:0.951 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:211-085-0
Documents
ACRO12035
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4535 to 1.4565 (20°C, 589 nm)
- GC:≥94 %
- Specific optical rotation:-5.5° to -7.9° (20°C, 589 nm) (c=1,CHCl3)
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



