MidiKros 41,5cm MPES 750kD 1,0mm 1/2TC X FLL dry 1 * 1 items
SPECT04-E750-10-N
MidiKros 41,5cm MPES 750kD 1,0mm 1/2TC X FLL dry 1 * 1 items
SPECT04-E750-10-N
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection