Order Entry
Italy
Orders LinkContactUsLinkComponent
Conductivity standards, Supelco®
Conductivity standards, Supelco®
Conductivity standard, 147 µmhos/cm, 1 L
Catalog # COND147-1L
Supplier:   Merck
Conductivity standards, Supelco®
Catalog # COND147-1L
Supplier:  Merck
CAS Number:  
Conductivity standards, Supelco®
Conductivity standard, 147 µmhos/cm, 1 L
CAS Number:  
Price
137,00 €
Log in for availability
1 * 1 L
Product Details & Documents
100 µmhos/cm = 100 µS/cm
Certifications: These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Documents
Specifications
Specifications
  • Pk:
    1 L
  • Description:
    Conductivity standard
  • Conductivity:
    147 µmhos/cm