LAL REAGENZ-WASSER, 100ML 1 * 30 Fl
Catalog # PYRQ4000100H-30
LAL REAGENZ-WASSER, 100ML 1 * 30 Fl
Catalog # PYRQ4000100H-30
Frequently Bought Together
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
ESD safety shoes, slip-on, X-Light, 7131032/7131037
Supplier: Abeba
Filter forceps
Supplier: Merck
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck