Spin columns, PAGE-Optimizer™
PAGE-Optimizer™, Spin column
Catalog # 786-948
Supplier: G-Biosciences
Jump to:
Product Details & Documents
PAGE-Optimizer™ is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents. PAGE-Optimizer™ helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer™ has also been shown to reveal protein bands previously masked by the above interfering agents.
- Eliminate smeared lanes
- Reveal key missing protein bands
- Resolve distorted bands for improved visualization
- Prevent overlapping lanes resulting in contamination and distortion ofadjacent lanes
- Reduce swollen lanes to prevent protein contamination
Specifications
- Type:Spin column
- Description:PAGE-Optimizer™