Order Entry
Italy
ContactUsLinkComponent
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperatur e sensor: Pt100, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 360 mm, Diameter: 12 1 * 1 items
Catalog # HAMI238632
Supplier:  HAMILTON BONADUZ
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperatur e sensor: Pt100, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 360 mm, Diameter: 12 1 * 1 items
Catalog # HAMI238632
Supplier:  HAMILTON BONADUZ
Frequently Bought Together