Order Entry
Italy
Orders LinkContactUsLinkComponent
CF® Dye dCTP conjugates

CF® Dye dCTP conjugates

CF®660R-dCTP, lyophilised, 25 Nmo
Catalog # 40068.
Supplier:   Biotium
CF® Dye dCTP conjugates
Catalog # 40068.
Supplier:  Biotium
CAS Number:  
CF® Dye dCTP conjugates

CF®660R-dCTP, lyophilised, 25 Nmo

CAS Number:  
Price
621,00 €
Log in for availability
1 * 25 Nmo
Product Details & Documents

CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.

  • Synthesise exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colours from green to far-red
  • CF® dyes have superior brightness, photostability, and are water-soluble

Caution: CF™594 dCTP is not compatible with PCR labelling of DNA probes.

Specifications

Specifications

  • Pk:
    25 Nmo
  • Description:
    CF®660R-dCTP, lyophilised
  • Cat. No.:
    40068.
  • Excitation max.:
    663 nm
  • Emission max.:
    682 nm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.