Drum sumps and pallets
Drum sump, PE, for 4×200 L drum, with polyethylene grid, 1220×1230×220 mm
Catalog # MSGA31862
Supplier: asecos
Jump to:
Product Details & Documents
This sump pallets are made of PE with varied capacities from 60 to 400 L.
- Compact and robust design
- Approved for the safe storage of water pollutants
- Swivel castors and handle for mobility
- Blue in colour
Sump pallets with PE grid or galvanised grid are helps in direct placing on the floor and provide resistant against oil, acid and alkali.
Feet present with sumps are ideal for in-house transport of the empty sump pallet.
Specifications
- Description:Drum sump, PE, for 4×200 L drum, with polyethylene grid
- Seq:12
- W×D×H:1220×1230×220 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



