NICERGOLINE FOR SYSTEM SUITABILITY 1 * 10 mg
Catalog # EDQMY0001352
Supplier: EDQM
Frequently Bought Together
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets
Supplier: Mettler - Toledo
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Calibration fluids and oils for refractometers
Supplier: BELLINGHAM STANLEY
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Methocarbamol, Reference Standard
Supplier: USP