Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Accessories for epMotion®
Accessories for epMotion®
Module reservoir rack, for 4× 5 ml Eppendorf tubes, temperable
Catalog # EPPE5075799.340
Supplier:   EPPENDORF
Accessories for epMotion®
Catalog # EPPE5075799.340
Supplier:  EPPENDORF
Accessories for epMotion®
Accessories for epMotion®
Module reservoir rack, for 4× 5 ml Eppendorf tubes, temperable
Price
400,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Descrizione:
    Module reservoir rack, for 4× 5 ml Eppendorf tubes, temperable
  • Seq:
    16