Flat bottom tubes, 112,5x16,1 mm, 15 ml, soda lime glass, with thread cap-alu 16, without screw cap, thickness of wall 0.95 mm, 299 pcs./tray, 1 * 1.196 items
Catalog # MARI3861400
Supplier: MARIENFELD
Flat bottom tubes, 112,5x16,1 mm, 15 ml, soda lime glass, with thread cap-alu 16, without screw cap, thickness of wall 0.95 mm, 299 pcs./tray, 1 * 1.196 items
Catalog # MARI3861400
Supplier: MARIENFELD
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection