Flasks, round bottom, with standard ground joints
100 ml, 24/29 NS, 110 mm, 64 mm
WITG0.602.100.E
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
- Ext.Ø:64 mm
- Socket size:24/29 NS
- Capacity:100 ml
- Height:110 mm
- Seq:15



