Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Flask, 250ml, ST29/32, pear shaped, outer diameter 85mm, height 150mm, borosilicate glass 3.3 1 * 1 items
Codice catalogo WITG0.664.250.E
Supplier:
witeg Labortechnik
Expand image
Flask, 250ml, ST29/32, pear shaped, outer diameter 85mm, height 150mm, borosilicate glass 3.3 1 * 1 items
Codice catalogo WITG0.664.250.E
Supplier:
witeg Labortechnik
Frequently Bought Together
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Earmuffs, K2
Supplier: UVEX
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection