Order Entry
Italy
ContactUsLinkComponent
Flask, 10ml, ST19/26, pear shaped, outer diameter 33mm, height 75mm, borosilicate glass 3.3 1 * 1 items
Codice catalogo WITG0.662.010.E
Supplier:  witeg Labortechnik
Flask, 10ml, ST19/26, pear shaped, outer diameter 33mm, height 75mm, borosilicate glass 3.3 1 * 1 items
Codice catalogo WITG0.662.010.E
Supplier:  witeg Labortechnik
Frequently Bought Together