Viscosity Standards(cSt) reads 41000@20C, 25000@25C, [email protected], 6700@40C, 3200@50C 1 * 500 mL
REGEREVIS-S8000
: REAGECON
Viscosity Standards(cSt) reads 41000@20C, 25000@25C, [email protected], 6700@40C, 3200@50C 1 * 500 mL
REGEREVIS-S8000
: REAGECON
Frequently Bought Together
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffers, ready to use
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo