Viscosity Standards(cSt) reads 59@20C, 47@25C, [email protected], 25@40C, 18@50C 1 * 500 mL
Catalog # REGEREVIS-N26
Supplier: REAGECON
Viscosity Standards(cSt) reads 59@20C, 47@25C, [email protected], 25@40C, 18@50C 1 * 500 mL
Catalog # REGEREVIS-N26
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW