Order Entry
Italy
ContactUsLinkComponent
Viscosity Standards(cSt) reads 10@20C, 8.7@25C, [email protected], 5.7@40C, 4.4@50C 1 * 500 mL
Catalog # REGEREVIS-S6
Supplier:  REAGECON
Viscosity Standards(cSt) reads 10@20C, 8.7@25C, [email protected], 5.7@40C, 4.4@50C 1 * 500 mL
Catalog # REGEREVIS-S6
Supplier:  REAGECON