Viscosity Standards(cSt) reads 110@20C, 87@25C, [email protected], 44@40C, 30@50C 1 * 500 mL
REGEREVIS-N44
: REAGECON
Viscosity Standards(cSt) reads 110@20C, 87@25C, [email protected], 44@40C, 30@50C 1 * 500 mL
REGEREVIS-N44
: REAGECON
Frequently Bought Together
Nitenpyram Analytical standard, reference material, Fluka™
: Honeywell Chemicals
in Acetonitrile 1 * 1 mL
: EHRENSTORFER
4'-Hydroxydiclofenac, Supelco®
: Merck
CEFAZOLIN SODIUM SALT 1 * 75 mg
: Merck
Nitrate standards for ISE calibration, Orion™
: Thermo Orion
Imazapyr certified 1 * 0,1 g
: EHRENSTORFER