Order Entry
Italy
ContactUsLinkComponent
The dNTP Mixes are ready-to-use aqueous solution at pH7.0 containing dATP, dCTP, dGTP and dTTP, each at a final concentration of 10 or 25mM. It has been functionally tested in PCR with Taq and Pfu DNA polymerases. The mix is Endo, exo-ribonuclease, a 1 * 1 mL
Catalog # GENO786-457
Supplier:  G-Biosciences
The dNTP Mixes are ready-to-use aqueous solution at pH7.0 containing dATP, dCTP, dGTP and dTTP, each at a final concentration of 10 or 25mM. It has been functionally tested in PCR with Taq and Pfu DNA polymerases. The mix is Endo, exo-ribonuclease, a 1 * 1 mL
Catalog # GENO786-457
Supplier:  G-Biosciences