Order Entry
Italy
Orders LinkContactUsLinkComponent
Sampling spoons, SteriPlast

Sampling spoons, SteriPlast

127 mm, 2,5 ml
Catalog # 231-0525
Supplier:   BURKLE
Sampling spoons, SteriPlast
Catalog # 231-0525
Supplier:  BURKLE
Sampling spoons, SteriPlast
Sampling spoons, SteriPlast

127 mm, 2,5 ml

Price
67,40 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.

  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed, gamma radiation sterilised
Specifications
  • Capacity:
    2,5 ml
  • Length:
    127 mm
  • Seq:
    01