Pneumatic lockouts, Quick-Disconnect
Red
BRDY065645
: Brady
These pneumatic quick-disconnect lockouts isolate pneumatic energy without the expense and inconvenience of installing in-line lockout valves.
- Device is fitted to the male fitting, isolating equipment from all compressed air sources
- Accommodates the majority of 6 mm, 9 mm and 12 mm male fittings
Max. shackle diameter: 7,84 mm
Min. shackle clearance: 20 mm
- Colour:Red
- Seq:02



