Order Entry
Italy
Orders LinkContactUsLinkComponent
CF® Dye dCTP conjugates

CF® Dye dCTP conjugates

CF®532-dCTP, lyophilised, 25 Nmo
Catalog # 40057.
Supplier:   Biotium
CF® Dye dCTP conjugates
Catalog # 40057.
Supplier:  Biotium
CAS Number:  
CF® Dye dCTP conjugates

CF®532-dCTP, lyophilised, 25 Nmo

CAS Number:  
Price
621,00 €
Log in for availability
1 * 25 Nmo
Product Details & Documents

CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.

  • Synthesise exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colours from green to far-red
  • CF® dyes have superior brightness, photostability, and are water-soluble

Caution: CF™594 dCTP is not compatible with PCR labelling of DNA probes.

Specifications

Specifications

  • Pk:
    25 Nmo
  • Description:
    CF®532-dCTP, lyophilised
  • Cat. No.:
    40057.
  • Excitation max.:
    527 nm
  • Emission max.:
    558 nm

Specifications