Order Entry
Italy
ContactUsLinkComponent
Microscope slides in tropical packing 76x26 mm, thickness 1 mm, white glass, Edges ground at 90, twin-frosted approx. 20mm, 50 pcs. paper-interleaved, 50 boxes in an aluminium foil bag, 2500 pcs./carton. 1 * 2.500 items
Catalog # MARI1000614
Supplier:  MARIENFELD
Microscope slides in tropical packing 76x26 mm, thickness 1 mm, white glass, Edges ground at 90, twin-frosted approx. 20mm, 50 pcs. paper-interleaved, 50 boxes in an aluminium foil bag, 2500 pcs./carton. 1 * 2.500 items
Catalog # MARI1000614
Supplier:  MARIENFELD